Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylimidazole, 99%

Catalog No. AC219280025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle

CAS: 1072-62-4 | C5H8N2 | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-62-4
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005192
InChI Key PQAMFDRRWURCFQ-UHFFFAOYSA-N
Synonym 2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972
PubChem CID 66130
IUPAC Name 2-ethyl-1H-imidazole
SMILES CCC1=NC=CN1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 83.0°C to 87.0°C
Boiling Point 268.0°C
Flash Point 159°C
Assay Percent Range 1% max. Imidazole (GC)
Packaging Plastic Bottle
Quantity 2.5 kg
Beilstein 23, 78
Solubility Information Solubility in water: 617g/L (20°C). Other solubilities: soluble in many organic solvents
Formula Weight 96.13
Percent Purity 99%
Chemical Name or Material 2-Ethylimidazole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 214-011-5
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.