Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethynylthiophene, ≥90%, Thermo Scientific™

Catalog No. p-3175177
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 4298-52-6
Molecular Formula C6H4S
Molecular Weight (g/mol) 108.16
MDL Number MFCD03425129
InChI Key LWISLHRIEATKTM-UHFFFAOYSA-N
Synonym 2-ethynyl-thiophene, 2-thienylethine, 2-thiophenylacetylene, 2-thienyl acetylene, pubchem16123, thien-2-yl acetylene, thiophene, 2-ethynyl, thiophen-2-yl acetylene, 2-ethynylthiophene, acmc-209js9
PubChem CID 5246258
IUPAC Name 2-ethynylthiophene
SMILES C#CC1=CC=CS1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 108.16
Percent Purity 90%
Chemical Name or Material 2-ethynylthiophene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.