Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-nitrotoluene, 98%

Catalog No. AAA1446414
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AAA1446414

Supplier: Thermo Scientific Chemicals A1446414

Only null left
Add to Cart

CAS: 1427-07-2 | C7H6FNO2 | 155.128 g/mol

It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.

Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1427-07-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00007199
InChI Key WIQISTBTOQNVCE-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrotoluene, toluene, 2-fluoro-4-nitro, benzene, 2-fluoro-1-methyl-4-nitro, 3-fluoro-4-methylnitrobenzene, 1-methyl-2-fluoro-4-nitrobenzene, 2-fluoro-1-methyl-4-nitro-benzene, pubchem1592, 2-fluoro4-nitrotoluene, acmc-209cov, 2-fluor-4-nitro-toluol
PubChem CID 74025
IUPAC Name 2-fluoro-1-methyl-4-nitrobenzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])F

Specifications

Melting Point 32°C to 37°C
Boiling Point 56°C to 58°C (1 mmHg)
Flash Point 73°C (163°F)
Quantity 25 g
UN Number UN2811
Beilstein 2250156
Solubility Information Insoluble in water. Solubility in methanol gives very faint turbidity.
Formula Weight 155.13
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-nitrotoluene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 215-845-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.