Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-methoxybenzoic acid, 97+%

Catalog No. p-7034673
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 367-83-9 | C8H7FO3 | 170.139 g/mol

2-Fluoro-5-methoxybenzoic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-methoxybenzoic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-83-9
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00272566
InChI Key REDSLTKMNCCQBC-UHFFFAOYSA-N
PubChem CID 2774543
IUPAC Name 2-fluoro-5-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)F)C(=O)O

Specifications

Quantity 250 mg
Synonym 2-fluoro-5-methoxy-benzoic acid, benzoic acid,2-fluoro-5-methoxy, benzoic acid, 2-fluoro-5-methoxy, pubchem4966, 6-fluoro-m-anisic acid, 3-carboxy-4-fluoroanisole, 2-fluoro-5-methoxybenzoicacid, 6-fluoro-3-methoxybenzoic acid, rarechem al bo 0748, buttpark 45\01-62
Formula Weight 170.14
Percent Purity ≥97%
Chemical Name or Material 2-Fluoro-5-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.