Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzylamine, 97%

Catalog No. AAA1562304
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1562304 2 g
AAA1562309 10 g
AAA1562318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1562304 Supplier Thermo Scientific Chemicals Supplier No. A1562304

CAS: 89-99-6 | C7H8FN | 125.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-99-6
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD00008107
InChI Key LRFWYBZWRQWZIM-UHFFFAOYSA-N
Synonym 2-fluorobenzylamine, 2-fluorophenyl methanamine, benzenemethanamine, 2-fluoro, o-fluorobenzylamine, benzylamine, o-fluoro, 1-2-fluorophenyl methanamine, 2-fluoro-benzylamine, 2-fluorobenzyl amine, 2-fluorophenyl methylamine, o-fluorobenzyl amine
PubChem CID 66649
IUPAC Name (2-fluorophenyl)methanamine
SMILES C1=CC=C(C(=C1)CN)F

Specifications

Density 1.099
Boiling Point 80°C to 82°C (18 mmHg)
Flash Point 67°C (152°F)
Odor Amine-like
Refractive Index 1.517
Quantity 2 g
UN Number UN2735
Beilstein 508109
Sensitivity Air Sensitive
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 2-Fluorobenzylamine
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 201-957-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.