Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluorocinnamic acid, predominantly trans, 98+%

Click to view available options
Quantity:
5g
25g
451-69-4
C9H7FO2
166.151
MFCD00004370
IOUDZAFBPDDAMK-AATRIKPKSA-N
2-fluorocinnamic acid,o-fluorocinnamic acid,cinnamic acid, o-fluoro,2-propenoic acid, 3-2-fluorophenyl,2e-3-2-fluorophenyl prop-2-enoic acid,3-2-fluorophenyl acrylic acid,3-2-fluoro-phenyl-acrylic acid,2e-3-2-fluorophenyl acrylic acid,3-2-fluorophenyl-2-propenoic acid,trans-2-fluorocinnamic acid
735833
(E)-3-(2-fluorophenyl)prop-2-enoic acid
C1=CC=C(C(=C1)C=CC(=O)O)F
451-69-4
C9H7FO2
166.151
MFCD00004370
IOUDZAFBPDDAMK-AATRIKPKSA-N
2-fluorocinnamic acid,o-fluorocinnamic acid,cinnamic acid, o-fluoro,2-propenoic acid, 3-2-fluorophenyl,2e-3-2-fluorophenyl prop-2-enoic acid,3-2-fluorophenyl acrylic acid,3-2-fluoro-phenyl-acrylic acid,2e-3-2-fluorophenyl acrylic acid,3-2-fluorophenyl-2-propenoic acid,trans-2-fluorocinnamic acid
735833
(E)-3-(2-fluorophenyl)prop-2-enoic acid
C1=CC=C(C(=C1)C=CC(=O)O)F
Melting Point 179°C to 183°C
Odor Odorless
Quantity 5g
Beilstein 1863963
Formula Weight 166.15
Percent Purity ≥98%
Chemical Name or Material 2-Fluorocinnamic acid, predominantly trans

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-195-3

TSCA : No

Recommended Storage : Ambient temperatures