Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furaldehyde, ACS, 98% min

Catalog No. p-7049925
Click to view available options
Quantity:
250 g
1 kg

CAS: 98-01-1 | C5H4O2 | 96.09 g/mol

It is primarily used in the production of furan derivatives such as furan and furfuryl alcohol. Another major application of furfural is as extraction solvent in refineries. It is also used to accelerate vulcanization, as an ingredient of phenolic resins (Durite) and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-01-1
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.09
MDL Number MFCD00003229
InChI Key HYBBIBNJHNGZAN-UHFFFAOYSA-N
Synonym furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
PubChem CID 7362
ChEBI CHEBI:34768
IUPAC Name furan-2-carbaldehyde
SMILES O=CC1=CC=CO1

Specifications

Melting Point -38°C
Color Yellow to Brown
Density 1.159 g/mL
Boiling Point 162°C
Flash Point 60°C (140°F)
Odor Almond-like
Refractive Index 1.5261
Quantity 250 g
UN Number UN1199
Beilstein 105755
Sensitivity Air Sensitive
Merck Index 14,4304
Solubility Information It is soluble in water.
Formula Weight 96.09
Percent Purity ≤98%
Grade ACS
Physical Form Liquid
Chemical Name or Material 2-Furaldehyde
Show More Show Less
Hazard Category H226-H301-H312-H315-H319-H330-H335-H373
Hazard Statement GHS H Statement
H301-H331-H226-H351-H312-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
Suspected of causing cancer.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: FURALDEHYDES
EINECSNumber 202-627-7
RTECSNumber LT7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.