Learn More
2-Furonitrile, 98+%
Description
2-Furonitrile is used as a extractive distillation solvent and as a sweetening agent. It is also used as an intermediate in pharmaceutical and fine chemical synthesis. 2-Furonitrile was employed as substrate to investigate the substrate specificity of nitrilase from Rhodococcus rhodochrous Jl cell.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Furonitrile is used as a extractive distillation solvent and as a sweetening agent. It is also used as an intermediate in pharmaceutical and fine chemical synthesis. 2-Furonitrile was employed as substrate to investigate the substrate specificity of nitrilase from Rhodococcus rhodochrous Jl cell.
Solubility
Soluble in most common organic solvents. Slightly soluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Specifications
Specifications
| Density | 1.062 |
| Boiling Point | 146°C to 148°C |
| Flash Point | 35°C (95°F) |
| Odor | Recognizable |
| Refractive Index | 1.479 |
| Quantity | 25 g |
| UN Number | UN1992 |
| Beilstein | 107033 |
| Solubility Information | Soluble in most common organic solvents. Slightly soluble in water. |
| Formula Weight | 93.09 |
| Show More |
Safety and Handling
| Hazard Category | H226-H302+H312+H332-H318 |
| Hazard Statement | GHS H Statement H311-H226-H302-H332 Toxic in contact with skin. Flammable liquid and vapor. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S. |
| EINECSNumber | 210-537-4 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.