Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-4-methoxybenzoic acid, 98%

Catalog No. p-7036889
Click to view available options
Quantity:
5 g
25 g

CAS: 2237-36-7 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2237-36-7
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002450
InChI Key MRIXVKKOHPQOFK-UHFFFAOYSA-N
Synonym 4-methoxysalicylic acid, 2-hydroxy-p-anisic acid, 2-hydroxy-4-methoxy-benzoic acid, benzoic acid, 2-hydroxy-4-methoxy, 2-hydroxy-4-methoxybenzoes, 4-methoxysalicylicacid, pubchem18895, 4-methoxy salicylic acid, 4-methoxy-salicylic acid, acmc-1cs6b
PubChem CID 75231
IUPAC Name 2-hydroxy-4-methoxybenzoic acid
SMILES COC1=CC=C(C(O)=O)C(O)=C1

Specifications

Melting Point 158°C to 159°C
Quantity 5 g
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-4-methoxybenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 218-801-0
RTECSNumber DH2270000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.