Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Hydroxyethyl)hydrazine

Catalog No. AA4379330
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Catalog No. Quantity
AA4379330 250 g
AA4379309 10 g
AA4379318 50 g
3 options

Catalog No. AA4379330

Supplier: Thermo Scientific Chemicals 04379330

Only null left
Add to Cart

CAS: 109-84-2 | C2H8N2O | 76.099 g/mol

(2-Hydroxyethyl)hydrazine is used as a plant growth regulant, a component in jet fuels, intermediate for organic synthesis especially heterocycles used in agrochemicals, pharmaceuticals, stabilizers and polymerizations, Chain extender for urethane formulations, corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-84-2
Molecular Formula C2H8N2O
Molecular Weight (g/mol) 76.099
MDL Number MFCD00007623
InChI Key GBHCABUWWQUMAJ-UHFFFAOYSA-N
Synonym 2-hydroxyethylhydrazine, 2-hydrazinoethanol, omaflora, ethanolhydrazine, 2-hydroxyethyl hydrazine, ethanol, 2-hydrazino, hydrazineethanol, 2-hydrazinoethyl alcohol, beta-hydroxyethylhydrazine, 2-hydrazineethanol
PubChem CID 8017
ChEBI CHEBI:50098
IUPAC Name 2-hydrazinylethanol
SMILES C(CO)NN

Specifications

Melting Point -70°C
Density 1.121 g/mL
Boiling Point 218°C to 220°C
Flash Point 73°C (163°F)
Refractive Index 1.493
Quantity 250 g
UN Number UN2810
Solubility Information Soluble in water,alcohols and glycols.
Formula Weight 76.1
Physical Form Liquid
Chemical Name or Material (2-Hydroxyethyl)hydrazine
Show More Show Less
Hazard Category H227-H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-711-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.