Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron trifluoride etherate, approx. 48% BF3

Catalog No. AC174560010
Click to view available options
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
Glass bottle

CAS: 109-63-7 | C4H10BF3O | 141.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-63-7
Molecular Formula C4H10BF3O
Molecular Weight (g/mol) 141.93
MDL Number MFCD00013194
InChI Key KZMGYPLQYOPHEL-UHFFFAOYSA-N
Synonym Boron trifluoride ethyl ether
PubChem CID 8000
IUPAC Name ethoxyethane;trifluoroborane
SMILES FB(F)F.CCOCC

Specifications

Melting Point -60.0°C
Color Colorless to Straw-Yellow
Density 1.1200g/mL
Boiling Point 126.0°C
Flash Point 58°C
Packaging Glass bottle
Refractive Index 1.3429 to 1.3449
Linear Formula (C2H5)2O·BF3
Quantity 1 kg
Fieser 01,70; 02,35; 03,33; 04,44; 05,52; 06,65; 07,31; 08,51; 09,64; 10,52; 11,72; 12,66; 13,3; 14,54; 15,45; 16,44; 17,52
Merck Index 15,1352
Specific Gravity 1.12
Solubility Information Solubility in water: reacts. Other solubilities: decomposes in alcohol,soluble in ether
Viscosity 1.89 mPa.s (20°C)
Formula Weight 141.93
Percent Purity ∼48% BF3
Physical Form Liquid, Fuming In Moist Air
Chemical Name or Material Boron trifluoride etherate, 48% BF3
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 203-689-8
TSCA TSCA
Recommended Storage Product darkens on storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.