Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxymethyl-12-crown-4, 97%

Catalog No. AA4450903
Click to view available options
Quantity:
0.25 g
1 g
5 g

CAS: 75507-26-5 | C9H18O5 | 206.238 g/mol

Ethylene oxide is important or critical to the production of detergents, thickeners, solvents, plastics, and various organic chemicals such as ethylene glycol, ethanolamines, simple and complex glycols, polyglycol ethers and other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75507-26-5
Molecular Formula C9H18O5
Molecular Weight (g/mol) 206.238
MDL Number MFCD00067524
InChI Key NJIPEIQHUNDGPY-UHFFFAOYSA-N
PubChem CID 2733085
IUPAC Name 1,4,7,10-tetraoxacyclododec-2-ylmethanol
SMILES C1COCCOC(COCCO1)CO

Specifications

Density 1.20 g/mL
Boiling Point 115°C (0.04 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.48
Quantity 1 g
Beilstein 4661968
Synonym 1,4,7,10-tetraoxacyclododecane-2-methanol, 2-hydroxymethyl-12-crown-4, 2-hydroxymethyl-12-crown4-ether, 1,4,7,10-tetraoxacyclododecan-2-ylmethanol, 2-hydroxymethyl-12-crown, 2-hydroxymethyl-12-crown 4-ether, 2-hydroxymethyl-12-crown-4-ether, 2-hydroxymethyl-1,2-crown 4-ether, 1,4,7,10-tetraoxacyclododecan-2-methanol, 2,5,8,11-tetraoxacyclododecyl methan-1-ol
Solubility Information Miscible in water. Soluble in non-polar solvents. Insoluble in water.
Formula Weight 206.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Hydroxymethyl-12-crown-4
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 278-224-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.