Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 2'-MeCCPA
GREENER_CHOICESDP

Catalog No. 228110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-811-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-811-0 Supplier Tocris Bioscience™ Supplier No. 2281/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Very selective and potent A1 receptor agonist

Potent and highly selective agonist at A1 adenosine receptors (Ki values are 3.3, 9580, 37600 and 1150 nM for human recombinant A1, A2A, A2B and A3 receptors respectively). Acts as a full agonist; inhibits forskolin-stimulated adenylyl cyclase activity in rat cortical membranes with an IC50 value of 13.1 nM.

Chemical Identifiers

CAS 205171-12-6
Molecular Formula C16H22ClN5O4

Specifications

Quantity 10 mg
Formula Weight 383.83
Purity >98%
Molecular Weight (g/mol) 383.833
Chemical Name or Material 2'-MeCCPA
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.