Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Acros Organics

Click to view available options
Quantity:
5g
10g
25g
19767-45-4
C2H5NaO3S2
164.18
XOGTZOOQQBDUSI-UHFFFAOYSA-M
mesna,sodium 2-mercaptoethanesulfonate,mesnex,uromitexan,mitexan,2-mercaptoethanesulfonic acid sodium salt,mistabron,mesnum,mistabronco,mucofluid
23662354
sodium;2-sulfanylethanesulfonate
C(CS(=O)(=O)[O-])S.[Na+]
19767-45-4
C2H5NaO3S2
164.18
XOGTZOOQQBDUSI-UHFFFAOYSA-M
mesna,sodium 2-mercaptoethanesulfonate,mesnex,uromitexan,mitexan,2-mercaptoethanesulfonic acid sodium salt,mistabron,mesnum,mistabronco,mucofluid
23662354
sodium;2-sulfanylethanesulfonate
C(CS(=O)(=O)[O-])S.[Na+]
Quantity 5g
Infrared Spectrum Authentic
Loss on Drying 2% max.
Assay Percent Range 97.5 to 102% min. (On dried substance) (Iodometry)
Merck Index 15,5979
Solubility Information Solubility in water: soluble.
Formula Weight 164.18
Percent Purity 98%
Chemical Name or Material 2-Mercaptoethanesulfonic acid sodium salt

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 243-285-9