Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-3-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. M21911G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 121643-44-5
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.13
MDL Number MFCD00153202
InChI Key SSAZZVQVJJXPMB-UHFFFAOYSA-N
Synonym 2-methoxy-3-trifluoromethyl pyridine, 2-methoxy-3-trifluoromethyl-pyridine, pyridine,2-methoxy-3-trifluoromethyl, pyridine, 2-methoxy-3-trifluoromethyl, pubchem2404, acmc-209aes, tpc-py090, 3-trifluoromethyl-2-methoxypyridine
PubChem CID 2775309
IUPAC Name 2-methoxy-3-(trifluoromethyl)pyridine
SMILES COC1=NC=CC=C1C(F)(F)F

Specifications

Color Colorless
Quantity 1 g
UN Number 1993
Formula Weight 177.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-3-(trifluoromethyl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.