Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxybutyl Acetate 97.0+%, TCI America™
SDP

Catalog No. A167525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A167525G 25 g
A16755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A167525G Supplier TCI America Supplier No. A167525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1173168-18-7
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.186
MDL Number MFCD06797047
InChI Key ZWUWDFWEMWMTHX-UHFFFAOYSA-N
Synonym Acetic Acid 2-Methoxybutyl Ester
PubChem CID 14993724
IUPAC Name 2-methoxybutyl acetate
SMILES CCC(COC(=O)C)OC

Specifications

Color Yellow
Quantity 25 g
Formula Weight 146.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxybutyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.