Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethanol, 99+%, for spectroscopy

Catalog No. p-156778
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 109-86-4 | C3H8O2 | 76.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.09
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Acidity 0.0008 meq/g max.
CAS Min % 99.0
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.05 max. at 270nm, 0.05 max. at 280nm, 0.1 max. at 260nm, 0.15 max. at 250nm, 0.2 max. at 240nm, 0.4 max. at 230nm
CAS Max % 100.0
Melting Point -85.0°C
Density 0.9600g/mL
Boiling Point 124.0°C
Flash Point 38°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.402
Linear Formula CH3OCH2CH2OH
Quantity 1 L
Residue after Evaporation 0.001% max.
Beilstein 01, 467
Specific Gravity 0.96
Merck Index 15, 6063
Solubility Information Solubility in water: soluble.
Viscosity 1.7 mPa.s (20°C)
Formula Weight 76.09
Percent Purity 99+%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May damage fertility.
May damage the unborn child.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/sh
EINECSNumber 203-713-7
RTECSNumber KL5775000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.