Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyhydroquinone, 99%, Thermo Scientific™

Catalog No. p-3736854
Click to view available options
Quantity:
50 g

Chemical Identifiers

CAS 824-46-4
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00013971
InChI Key LAQYHRQFABOIFD-UHFFFAOYSA-N
Synonym 2-methoxyhydroquinone, methoxyhydroquinone, 1,4-benzenediol, 2-methoxy, 1,4-dihydroxy-2-methoxybenzene, unii-2hi6hnr5u1, 2hi6hnr5u1, o-methoxyhydroquinone, 2,5-dihydroxyanisole, 2-mohydrop, acmc-209pp0
PubChem CID 69988
IUPAC Name 2-methoxybenzene-1,4-diol
SMILES COC1=C(C=CC(=C1)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 87°C to 90°C
Color Brown-Gray to Brown-Orange
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 50 g
Formula Weight 140.14
Percent Purity ≥98.5%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2-Methoxyhydroquinone
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.