Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1,3-dioxolane, 98%, Thermo Scientific™

Catalog No. AA2022806
Click to view available options
Quantity:
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-26-7
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00003210
InChI Key HTWIZMNMTWYQRN-UHFFFAOYSA-N
Synonym 1,3-dioxolane, 2-methyl, 2-methyldioxole, 2-methyldioxolane, methyldioxolane, methyl dioxolane, 2-methyl-1,3-dioxacyclopentane, dioxolane, methyl, unii-94w0r6v90a, acetaldehyde ethylene acetal, methyl-1,3-dioxolane
PubChem CID 10342
IUPAC Name 2-methyl-1,3-dioxolane
SMILES CC1OCCO1

Specifications

Density 0.982g/mL
Boiling Point 80°C to 83°C
Flash Point −2° (28°F)
Refractive Index 1.3980
Quantity 5 g
UN Number UN1993
Formula Weight 88.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methyl-1,3-dioxolane
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.