Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbenzofuran, 96%

Catalog No. p-4228784
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 4265-25-2 | C9H8O | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4265-25-2
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00005850
InChI Key GBGPVUAOTCNZPT-UHFFFAOYSA-N
Synonym 2-methylbenzofuran, benzofuran, 2-methyl, 2-methylcumarone, 2-methylbenzo b furan, 2-methyl benzofuran, unii-8x3183bz3x, methylbenzofuran, methyl benzoxole, 2-methyl-benzofuran, #
PubChem CID 20263
IUPAC Name 2-methyl-1-benzofuran
SMILES CC1=CC2=CC=CC=C2O1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0570g/mL
Boiling Point 197.0°C to 198.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5618
Quantity 1 g
Beilstein 17,6
Specific Gravity 1.057
Formula Weight 132.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Methylbenzofuran
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 224-249-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.