Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbutane, BAKER™, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Aluminum Bottle

Chemical Identifiers

78-78-4
C5H12
72.151
QWTDNUCVQCZILF-UHFFFAOYSA-N
6556
CHEBI:30362
2-methylbutane
CCC(C)C

A grade of chemicals for laboratory use

Packaging Aluminum Bottle
Specific Gravity 1L = 0.62kg
Flash Point −51.00°C
Quantity 4L
Formula Weight 72.15
Chemical Name or Material 2-Methylbutane