Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylindoline 98.0+%, TCI America™
SDP

Catalog No. m087125ml Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL

Chemical Identifiers

CAS 6872-06-6
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00005706
InChI Key QRWRJDVVXAXGBT-UHFFFAOYNA-N
PubChem CID 23305
IUPAC Name 2-methyl-2,3-dihydro-1H-indole
SMILES CC1CC2=CC=CC=C2N1

Specifications

Color Yellow
Boiling Point 230°C
Quantity 25 mL
Formula Weight 133.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylindoline
EINECSNumber (5)-0141
RTECSNumber NM1926350
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.