Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthylacetonitrile 98.0+%, TCI America™
SDP

Catalog No. N087425G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 7498-57-9
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004122
InChI Key LPCWDVLDJVZIHA-UHFFFAOYSA-N
Synonym 2-naphthylacetonitrile, 2-naphthalen-2-yl acetonitrile, 2-naphthaleneacetonitrile, 2-naphthyl acetonitrile, naphthalene-2-acetonitrile, 2-2-naphthyl acetonitrile, .beta.-naphthyleneacetonitrile, 2-naphtylacetonitrile, 2-napthylacetonitrile, 2-naphthylmethylcyanide
PubChem CID 82008
IUPAC Name 2-(naphthalen-2-yl)acetonitrile
SMILES N#CCC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 86°C
Color White-Yellow
Boiling Point 303°C
Quantity 25 g
UN Number 3439
Formula Weight 167.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthylacetonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.