Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzenesulfonic Acid Hydrate 98.0+%, TCI America™
SDP

Catalog No. N013925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N013925G 25 g
N01395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N013925G Supplier TCI America Supplier No. N013925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 80-82-0
Molecular Formula C6H5NO5S
Molecular Weight (g/mol) 203.168
MDL Number MFCD00053617
InChI Key HWTDMFJYBAURQR-UHFFFAOYSA-N
PubChem CID 6664
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)O

Specifications

Color Yellow
Quantity 25 g
UN Number 2305
Formula Weight 203.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzenesulfonic Acid Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.