Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzyl Cyanide 98.0+%, TCI America™
SDP

Catalog No. N078125G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. N078125G

Supplier: TCI America N078125G

Chemical Identifiers

CAS 610-66-2
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00007183
InChI Key YPRFCQAWSNWRLM-UHFFFAOYSA-N
Synonym 2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl
PubChem CID 11888
IUPAC Name 2-(2-nitrophenyl)acetonitrile
SMILES [O-][N+](=O)C1=CC=CC=C1CC#N

Specifications

Melting Point 84°C
Boiling Point 178°C
Quantity 25 g
UN Number 3439
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzyl Cyanide
EINECSNumber (3)-1820
RTECSNumber AM1248000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.