Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phthalimidopropionic acid, 98%, Thermo Scientific™

Catalog No. AAL1353403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1353403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1353403 Supplier Thermo Scientific Chemicals Supplier No. L1353403

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19506-87-7
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
MDL Number MFCD00023061
InChI Key OZWUITKBAWTEAQ-UHFFFAOYSA-N
Synonym 2-phthalimidopropionic acid, phthaloyl-dl-alanine, 2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 2-1,3-dioxoisoindolin-2-yl propanoic acid, phthalyl-dl-alanine, 2-1,3-dioxoisoindol-2-yl propanoic acid, 2-1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl propanoic acid, 2-phthalimidopropanoic acid, 2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, 2h-isoindole-2-acetic acid, 1,3-dihydro-alpha-methyl-1,3-dioxo
PubChem CID 86884
IUPAC Name 2-(1,3-dioxoisoindol-2-yl)propanoic acid
SMILES CC(C(=O)O)N1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 162°C to 164°C
Quantity 1 g
Beilstein 86452
Formula Weight 219.2
Percent Purity 98%
Chemical Name or Material 2-Phthalimidopropionic acid
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 243-117-4
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.