Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propene-1-sulfonic acid sodium salt, 95%, pract.

Catalog No. ac418370250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

CAS: 2495-39-8 | C3H5NaO3S | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2495-39-8
Molecular Formula C3H5NaO3S
Molecular Weight (g/mol) 144.13
MDL Number MFCD00051416
InChI Key DIKJULDDNQFCJG-UHFFFAOYSA-M
Synonym sodium allylsulfonate, sodium prop-2-ene-1-sulfonate, 2-propene-1-sulfonic acid, sodium salt, sodium allyl sulfonate, sodium 2-propene-1-sulfonate, unii-7sgf7tb9o2, sodium prop-2-enesulphonate, allylsulfonic acid sodium salt, sodium allysulfonate, 2-propene-1-sulfonic acid sodium salt
PubChem CID 23690996
IUPAC Name sodium;prop-2-ene-1-sulfonate
SMILES C=CCS(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH2CHCH2SO3Na
Quantity 25 g
Solubility Information Solubility in water: 1g/25mL. Other solubilities: soluble in alcohol
Formula Weight 144.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Propene-1-sulfonic acid sodium salt, pract., 95%
Show More Show Less
EINECSNumber 219-676-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.