Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propoxyethylamine 97.0+%, TCI America™
SDP

Catalog No. A132525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A132525ML 25 mL
A13255ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A132525ML Supplier TCI America Supplier No. A132525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42185-03-5
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00047955
InChI Key HMWXCSCBUXKXSA-UHFFFAOYSA-N
Synonym 2-Aminoethyl Propyl Ether, O-Propylethanolamine
PubChem CID 111878
IUPAC Name 2-propoxyethanamine
SMILES CCCOCCN

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 mL
UN Number 2733
Formula Weight 103.17
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Propoxyethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.