Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(Trifluoromethoxy)benzylamine 98.0+%, TCI America™

Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Color | Yellow |
Boiling Point | 77°C |
Quantity | 5 g |
UN Number | 2735 |
Formula Weight | 191.15 |
Percent Purity | ≥98.0% (GC,T) |
Physical Form | Liquid |
Chemical Name or Material | 2-(Trifluoromethoxy)benzylamine |
Safety and Handling
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction