Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

25,27-Dipropoxycalix[4]arene, 95%, Thermo Scientific Chemicals

Catalog No. AC327412500
Click to view available options
Quantity:
250 mg

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162301-48-6
Molecular Formula C34H36O4
Molecular Weight (g/mol) 508.658
MDL Number MFCD01632729
InChI Key JJYQEKZDWWUIQB-UHFFFAOYSA-N
Synonym 25,27-dipropoxycalix 4 arene, 2,6'-2-propoxy-1,3-phenylenebismethylene-2',6-2-propoxy-1,3-phenylenebismethylene diphenol
PubChem CID 2733466
SMILES CCCOC1=C2CC3=C(C(=CC=C3)CC4=C(C(=CC=C4)CC5=CC=CC(=C5O)CC1=CC=C2)OCCC)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 508.66
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material 25, 27-Dipropoxycalix[4]arene, 95%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.