Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GVS AcetatePlus™ 25 mm Cellulose Acetate Disc
Shop All Osmonics Products
Click to view available options
Pore Size:
0.45 μm
0.65 μm
5 μm
Description
Properties
- Membranes are composed of pure cellulose acetate that is internally supported by an inert polyester web. This web gives each membrane exceptional strength to prevent cracking, tearing, breaking and distortion when handled or creased.
- The resulting membrane has dimensional stability that can withstand autoclaving or steam sterilizing leaving the membrane unaffected in temperatures up to 274°F (135°C).
- The exceptional dimensional strength and low binding characteristics of the cellulose acetate filtration membranes provides higher throughputs than competitive offerings and reduces the amount of filter changes needed during proteinaceous solution filtering.
Characteristics
- Superior strength can withstand aggressive handling or be used with automated equipment without breaking or tearing
- Low extractables ensures tests will be clean with consistent results
- Hydrophilic wets out rapidly.
- Lot-to-lot consistency quality checks ensure consistent flow and diffusion rates for dependable results every time.
- Nonlysing of cells prevents contamination of critical solutions.
- Pore size range: 0.1 μm to 20 μm
Applications
- Biological fluid sterilization
- Cold sterilization
- Protein and enzyme filtration
- Tissue culture media sterilization
Specifications
Specifications
| Autoclavable | Autoclavable |
| Disposable | Limited-use |
| Filter Type | Membrane Filter |
| Format | Disc |
| Material | Cellulose Acetate |
| Pore Size | 0.65 μm |
| Sterility | Non-sterile |
| Type | Disc Membrane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction