Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

25-OH Vitamin D3 Antibody-antigen Complex Antibody (RM428), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB293941 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293941 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293941 Supplier Novus Biologicals Supplier No. NBP326039AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

25-OH Vitamin D3 Antibody-antigen Complex Monoclonal antibody specifically detects 25-OH Vitamin D3 Antibody-antigen Complex in Nonspecies specific samples. It is validated for ELISA

Specifications

Antigen 25-OH Vitamin D3 Antibody-antigen Complex
Applications ELISA
Classification Monoclonal
Clone RM428
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen 25-OH Vitamin D3 Antibody-antigen Complex
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.