Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

26S proteasome subunit 9 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB144191 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB144191 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB144191 Supplier Novus Biologicals Supplier No. NBP305933AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

26S proteasome subunit 9 Polyclonal specifically detects 26S proteasome subunit 9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen 26S proteasome subunit 9
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 26S proteasome non-ATPase regulatory subunit 9, p27, proteasome (prosome, macropain) 26S subunit, non-ATPase, 9, Rpn4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human 26S proteasome subunit 9 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5715
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.