Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2A Peptide Antibody (3H4) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013620 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013620 Supplier Novus Biologicals Supplier No. NBP280547
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

2A Peptide Monoclonal specifically detects 2A Peptide in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen 2A Peptide
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 3H4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1 μg/mL, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias 2A cleavage peptide, 2A peptide, 2A peptide tag, 2A sequence, Anti- MDV 2A-peptide, anti-2A peptide, anti-2A sequence, anti-2A-like peptide, antibody for 2A-containing recombinant proteins, anti-P2A tagged proteins, anti-P2A peptide, anti-T2A tagged proteins, anti-T2A peptide, anti-Thosea asigna virus 2A, FMDV 2A-peptide, peptide 2A sequences, self-cleaving'2A peptide2a pept, Thosea asigna virus 2A
Host Species Mouse
Immunogen KLH-coupled synthetic peptide corresponding to 2A peptide of Thosea asigna virus (CGDVEENPG)
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.