Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2A Peptide Antibody (3H4), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP259627V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259627V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259627V Supplier Novus Biologicals Supplier No. NBP259627V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

2A Peptide Monoclonal specifically detects 2A Peptide in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen 2A Peptide
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3H4
Conjugate DyLight 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2A cleavage peptide, 2A peptide, 2A peptide tag, 2A sequence, Anti- MDV 2A-peptide, anti-2A peptide, anti-2A sequence, anti-2A-like peptide, antibody for 2A-containing recombinant proteins, anti-P2A tagged proteins, anti-P2A peptide, anti-T2A tagged proteins, anti-T2A peptide, anti-Thosea asigna virus 2A, FMDV 2A-peptide peptide 2A sequences, self-cleaving'2A peptide2a pept, Thosea asigna virus 2A
Host Species Mouse
Immunogen KLH-coupled synthetic peptide corresponding to 2A peptide of Thosea asigna virus (CGDVEENPG)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This 2A peptide antibody is specific for T2A and P2A tagged proteins. Specificity to F2A and E2A proteins has not been tested.
Target Species Non-species specific, Other
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.