Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2A Peptide Antibody (3H4R), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB287584 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287584 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287584 Supplier Novus Biologicals Supplier No. NBP309171V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

2A Peptide Monoclonal antibody specifically detects 2A Peptide in Human, Nonspecies specific samples. It is validated for Western Blot

Specifications

Antigen 2A Peptide
Applications Western Blot
Classification Monoclonal
Clone 3H4R
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 2A cleavage peptide, 2A Peptide, 2A peptide tag, 2A sequence, 2A-like peptide, antibody for 2A-containing recombinant proteins, FMDV 2A-peptide, MDV 2A-peptide, P2A peptide, P2A tagged proteins, peptide 2A sequences, self-cleaving 2A peptide, T2A peptide, T2A tagged proteins, Thosea asigna virus 2A
Host Species Mouse
Immunogen This Recombinant 2A Peptide Antibody (3H4) was developed against KLH-coupled synthetic peptide corresponding to 2A peptide of Thosea asigna virus (CGDVEENPG)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Human, Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.