Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2B4/CD244/SLAMF4 Antibody (146506), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB003043 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003043 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003043 Supplier Novus Biologicals Supplier No. MAB10392IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen 2B4/CD244/SLAMF4
Applications Flow Cytometry
Classification Monoclonal
Clone 146506
Conjugate DyLight 755
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL
Gene Symbols CD244
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived partial recombinant human 2B4/CD244/SLAMF4 protein (amino acids 22-221)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Myeloid derived Suppressor Cell, Phospho Specific
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.