Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2B4/CD244/SLAMF4 Antibody (244F4), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP20223646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP20223646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP20223646 Supplier Novus Biologicals Supplier No. NBP200223JF646
Add to Cart
Add to Cart

Rat Monoclonal Antibody

2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen 2B4/CD244/SLAMF4
Applications Flow Cytometry
Classification Monoclonal
Clone 244F4
Conjugate Janelia Fluor 646
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL
Gene Symbols CD244
Host Species Rat
Immunogen The immunogen for this antibody is CD244.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 51744
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.