Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2B4/CD244/SLAMF4 Antibody (2B4.69), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N106589APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106589APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106589APC7 Supplier Novus Biologicals Supplier No. NB10065889APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen 2B4/CD244/SLAMF4
Applications Flow Cytometry
Classification Monoclonal
Clone 2B4.69
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL
Gene Symbols CD244
Host Species Mouse
Immunogen CD244 Fusion protein.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Myeloid derived Suppressor Cell, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 51744.0
Test Specificity NB100-65889 is specific for the CD244 cell surface antigen, also known as natural killer receptor 2B4 (NKG2B4). The glycoprotein CD244 is a member of the SLAM family of receptors which is expressed on natural killer (NK) cells, gamma delta T cells, CD14+ monocytes, and a subset of CD8+ T cells. CD244 was originally identified as an activating receptor on NK cells, although recent studies suggest that the receptor may have a wide range of activities including NK cell activation, costimulation, or even inhibition, depending on the cell type and stage of differentiation. The ligand of 2B4 is CD48, a GPI-anchored membrane protein also belonging to the SLAM family.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.