Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2B4/CD244/SLAMF4 Antibody (999602) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB401626 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401626 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401626 Supplier Novus Biologicals Supplier No. FAB10393J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

2B4/CD244/SLAMF4 Monoclonal antibody specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF

Specifications

Antigen 2B4/CD244/SLAMF4
Applications Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Classification Monoclonal
Clone 999602
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human 2B4/CD244/SLAMF4, Gly19-His222, Accession # NP_057466
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Myeloid derived Suppressor Cell, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 51744
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.