Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2B4/CD244/SLAMF4 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB029828 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB029828 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB029828 Supplier Novus Biologicals Supplier No. NBP176558AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

2B4/CD244/SLAMF4 Polyclonal specifically detects 2B4/CD244/SLAMF4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen 2B4/CD244/SLAMF4
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human SLAMF4. The immunogen is located within the last 50 amino acids of SLAMF4. Amino Acid Squence: PKAQNPARLSRKELEN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Myeloid derived Suppressor Cell, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 51744
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.