Learn More
2B4/CD244/SLAMF4 Antibody (MM0070-7D24), Alexa Fluor™ 532, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | 2B4/CD244/SLAMF4 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | MM0070-7D24 |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL |
| Gene Symbols | CD244 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.