Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2H-Chromene-3-carboxylic acid, 90+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
Specifications
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 176.17 |
Percent Purity | 90% |
Chemical Name or Material | 2H-Chromene-3-carboxylic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction