Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R)-(-)-Glycidyl tosylate, 99+%, Thermo Scientific™

Catalog No. AC264590050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113826-06-5
Molecular Formula C10H12O4S
Molecular Weight (g/mol) 228.262
MDL Number MFCD00010834
InChI Key NOQXXYIGRPAZJC-SECBINFHSA-N
Synonym 2r---glycidyl tosylate, r-glycidyl tosylate, r---glycidyltosylate, --glycidyl tosylate, 2r---glycidyltosylate, r---glycidyl tosylate, r-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6399, 2r-glycidyl tosylate, 2r-oxiran-2-ylmethyl 4-methylbenzenesulfonate
PubChem CID 154187
IUPAC Name [(2R)-oxiran-2-yl]methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 45.0°C to 48.0°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition − 18.00 (20.00°C c=2.5,CHCl3)
Specific Rotation − 18.00
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in chloroform
Formula Weight 228.27
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material (2R)-(-)-Glycidyl tosylate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Suspected of causing cancer.
May cause an allergic skin reaction.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.