Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ 2XYT-Broth, Capsules: Regular
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
454 g
1000 g
5000 g
Format:
Capsule
Description
Regular size capsules.
- Contents/L: 16g tryptone, 10g yeast extract, 5g NaCl
- Use 31 capsules/L

Specifications
Specifications
Certifications/Compliance | Molecular Biology Certified™ |
Content And Storage | 15°C to 30°C |
Format | Capsule |
Product Type | Microbiology Media |
Formulation | Tryptone 16g/L, Yeast extract 10g/L, NaCl 5g/L |
For Use With (Application) | For growth and maintenance of M13 phage or other filamentous phage |
Quantity | 1000 g |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction