Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ 2XYT Broth (Granulated Media)

Catalog No. p-4917392 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging Type:
Poly Bottle
Poly Pail

Used in the growth and cultivation of microorganisms in the lab and include E. coli, bacteriophage, and other bacterial strains

  • Granulated Media
  • Formulation per liter: Casein Peptone [91079-40-2] 16g, Yeast Extract [8013-01-2] 10g, Sodium Chloride [7647-14-5] 5g
  • Store at room temperature

TRUSTED_SUSTAINABILITY

Specifications

Form Solid
Media Type Dehydrated 2XYT
Product Type Microbiology Media
Formulation Casein Peptone 16g/L, Yeast extract 10g/L, Sodium Chloride 5g/L
Packaging Type Poly Pail
Quantity 2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.