Learn More
3,3',5,5'-Tetramethylbenzidine, MP Biomedicals™
Catalog No. p-7236587
Click to view available options
Quantity:
100 mg
1 g
5 g
3,3',5,5'-Tetramethylbenzidine is suitable as peroxidase substrate for use in ELISA procedures.
  • The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620-650 nm.
  • The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm.
  • A sensitive and specific reagent for the detection of blood, assay of hemoglobin, assay of peroxidases.
  • It is useful in gel staining procedure for low levels of home-associated peroxidases and for enzyme immunoassay of horseradish peroxidase.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mg
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.