Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethyl-1-butene, 95%

Catalog No. p-116359
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Catalog No. AC152720500

Supplier: Thermo Scientific Chemicals 152720500

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 558-37-2 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 558-37-2
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00008853
InChI Key PKXHXOTZMFCXSH-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-butene, neohexene, 1-butene, 3,3-dimethyl, tert-butylethylene, tert-butylethene, tert-hexene, trimethylvinylmethane, 3,3-dimethylbutene, 2,2-dimethyl-3-butene, unii-01zb73d2kk
PubChem CID 11210
IUPAC Name 3,3-dimethylbut-1-ene
SMILES CC(C)(C)C=C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -115.0°C
Color Colorless
Density 0.6500g/mL
Boiling Point 41.0°C
Flash Point −28°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCH=CH2
Refractive Index 1.3750 to 1.3780
Quantity 50 mL
Beilstein 01, 217
Specific Gravity 0.65
Formula Weight 84.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-1-butene, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 209-195-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.