Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethylpentane, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
562-49-2
C7H16
100.2
MFCD00009322
AEXMKKGTQYQZCS-UHFFFAOYSA-N
pentane, 3,3-dimethyl,pentane,3,3-dimethyl,unii-r1rh0vbd2l,r1rh0vbd2l,3,3,-dimethylpentane,acmc-1b14l,dsstox_cid_29275,dsstox_rid_83394,dsstox_gsid_49319,3,3-dimethylpentane
11229
3,3-dimethylpentane
CCC(C)(C)CC
562-49-2
C7H16
100.2
MFCD00009322
AEXMKKGTQYQZCS-UHFFFAOYSA-N
pentane, 3,3-dimethyl,pentane,3,3-dimethyl,unii-r1rh0vbd2l,r1rh0vbd2l,3,3,-dimethylpentane,acmc-1b14l,dsstox_cid_29275,dsstox_rid_83394,dsstox_gsid_49319,3,3-dimethylpentane
11229
3,3-dimethylpentane
CCC(C)(C)CC
Color Colorless
Specific Gravity 0.69
Refractive Index 1.3900 to 1.392
Packaging Glass Bottle
Flash Point -6°C
Boiling Point 86°C
Melting Point -135°C
Density 0.6900g/mL
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C2H5C(CH3)2C2H5
Beilstein 01,158
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether,miscible with acetone
Formula Weight 100.2
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 3, 3-Dimethylpentane
Show More Show Less

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger