Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-7-ylboronic acid, 90+%, Thermo Scientific™

Catalog No. CC13212DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13212DA 1 g Amber Glass Bottle
CC13212CB 250 mg Amber Glass Bottle
CC13212DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC13212DA Supplier Thermo Scientific Chemicals Supplier No. CC13212DA
Add to Cart

Chemical Identifiers

CAS 279261-89-1
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02681910
InChI Key CSNCBPVLUIFJOS-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl boronic acid, 3,4-dihydro-2h-1,5-benzodioxepine-7-boronic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-yl boronic acid, boronic acid,b-3,4-dihydro-2h-1,5-benzodioxepin-7-yl, 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-ylboronic acid, boronic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-yl, pubchem7845, acmc-1crzt, 3,4-dihydro-2h-1,5-benzodioxepin-7-boronic acid
PubChem CID 2776398
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepin-7-ylboronic acid
SMILES OB(O)C1=CC=C2OCCCOC2=C1

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 193.99
Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepin-7-ylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.